Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hot-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073405.2 | NP_505806.1 | 4 - 5 | 423 | 71 | |
GenBank mRNA | AF324058.1 | - | 4 - 5 | 360 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |