Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pptr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129373.4 | NP_001122845.1 | 3 - 4 | 305 | 91 | |
NM_001269354.3 | NP_001256283.1 | 3 - 4 | 206 | 91 | ||
NM_001383395.1 | 4 - 5 | 509 | 91 | |||
NM_001383396.1 | 3 - 4 | 339 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |