Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pptr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129373.4 | NP_001122845.1 | 5 - 6 | 1682 | 85 | |
NM_001269354.3 | NP_001256283.1 | 5 - 6 | 1583 | 85 | ||
NM_001383395.1 | 6 - 7 | 1886 | 85 | |||
NM_001383396.1 | 5 - 6 | 1716 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |