Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073412.2 | NP_505813.1 | 2 - 3 | 393 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |