Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: math-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028841.5 | NP_001024012.1 | 4 - 5 | 2114 | 112 | |
NM_001269365.3 | NP_001256294.1 | 4 - 5 | 2113 | 112 | ||
NM_001269366.3 | NP_001256295.1 | 4 - 5 | 2113 | 112 | ||
NM_001269367.3 | NP_001256296.1 | 4 - 5 | 2114 | 112 | ||
NM_001269368.3 | NP_001256297.1 | 4 - 5 | 2114 | 112 | ||
NM_001269369.3 | NP_001256298.1 | 4 - 5 | 2109 | 112 | ||
NM_001307753.4 | NP_001294682.1 | 4 - 5 | 2123 | 112 | ||
NM_001307754.4 | NP_001294683.1 | 4 - 5 | 2122 | 112 | ||
NM_001307755.3 | NP_001294684.1 | 4 - 5 | 2122 | 112 | ||
NM_001307756.3 | NP_001294685.1 | 4 - 5 | 2116 | 112 | ||
NM_001307757.3 | NP_001294686.1 | 4 - 5 | 2122 | 112 | ||
NM_001307758.3 | NP_001294687.1 | 4 - 5 | 2122 | 112 | ||
NM_001307759.4 | NP_001294688.1 | 4 - 5 | 2097 | 112 | ||
NM_001307760.4 | NP_001294689.1 | 4 - 5 | 2118 | 112 | ||
NM_073424.4 | NP_505825.1 | 4 - 5 | 2107 | 112 | ||
NM_073425.8 | NP_505826.1 | 4 - 5 | 2113 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |