Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380757.1 | NP_001367200.1 | 3 - 4 | 325 | 116 | |
NM_001380758.2 | NP_001367199.1 | 3 - 4 | 338 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |