Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129546.4 | NP_001123018.1 | 1 - 2 | 148 | 137 | |
NM_073439.3 | NP_505840.1 | 3 - 4 | 422 | 137 | ||
GenBank mRNA | AY008124.1 | - | 3 - 4 | 304 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |