Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073474.6 | NP_505875.2 | 6 - 7 | 1776 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |