Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073491.2 | NP_505892.1 | 1 - 2 | 307 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |