Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccch-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073525.8 | NP_505926.2 | 6 - 7 | 863 | 107 | |
NM_073526.5 | NP_505927.2 | 4 - 5 | 908 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |