Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380769.2 | NP_001366648.1 | 3 - 4 | 532 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |