Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269419.3 | NP_001256348.1 | 4 - 5 | 462 | 78 | |
NM_001269420.3 | NP_001256349.1 | 1 - 2 | 146 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |