Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313082.3 | NP_001300011.1 | 4 - 5 | 524 | 102 | |
NM_073559.8 | NP_505960.3 | 9 - 10 | 1546 | 102 | ||
NR_132532.1 | 8 - 9 | 1178 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |