Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073588.5 | NP_505989.1 | 12 - 13 | 2177 | 87 | |
GenBank mRNA | AY887910.1 | - | 12 - 13 | 2199 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |