Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073588.5 | NP_505989.1 | 6 - 7 | 1427 | 151 | |
GenBank mRNA | AY887910.1 | - | 6 - 7 | 1449 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |