Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073589.5 | NP_505990.3 | 11 - 12 | 1864 | 97 | |
GenBank mRNA | AY887911.1 | - | 11 - 12 | 1886 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |