Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073589.5 | NP_505990.3 | 5 - 6 | 790 | 124 | |
GenBank mRNA | AY887911.1 | - | 5 - 6 | 812 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |