Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073621.5 | NP_506022.1 | 7 - 8 | 1839 | 67 | |
GenBank mRNA | AF319616.1 | - | 7 - 8 | 1861 | 67 | |
AF328908.1 | - | 7 - 8 | 1870 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |