Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269479.4 | NP_001256408.1 | 6 - 7 | 785 | 70 | |
NR_069690.1 | 5 - 6 | 1082 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |