Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073671.6 | NP_506072.2 | 6 - 7 | 1076 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |