Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073672.6 | NP_506073.2 | 7 - 8 | 1233 | 115 |
Gene Symbol: T04H1.12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380781.1 | NP_001368636.1 | 8 - 9 | 1515 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |