Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073684.5 | NP_506085.2 | 1 - 2 | 279 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |