Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129512.4 | NP_001122984.1 | 3 - 4 | 710 | 92 | |
NM_001276939.4 | NP_001263868.1 | 5 - 6 | 465 | 92 | ||
NM_001276940.3 | NP_001263869.1 | 4 - 5 | 308 | 92 | ||
NM_073690.10 | NP_506091.2 | 3 - 4 | 239 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |