Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129512.4 | NP_001122984.1 | 8 - 9 | 1517 | 99 | |
NM_001276939.4 | NP_001263868.1 | 10 - 11 | 1272 | 99 | ||
NM_001276940.3 | NP_001263869.1 | 9 - 10 | 1115 | 99 | ||
NM_073690.10 | NP_506091.2 | 8 - 9 | 1046 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |