Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276941.4 | NP_001263870.1 | 3 - 4 | 356 | 145 | |
NM_001380784.1 | NP_001367281.1 | 3 - 4 | 321 | 145 | ||
NM_001380785.1 | NP_001366791.1 | 3 - 4 | 334 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |