Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073709.6 | NP_506110.1 | 4 - 5 | 434 | 92 | |
GenBank mRNA | AY728064.1 | - | 3 - 4 | 405 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |