Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047710.1 | NP_001041175.1 | 3 - 4 | 422 | 112 | |
NM_001322589.1 | NP_001309669.1 | 1 - 2 | 299 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |