Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ril-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028487.7 | NP_001023658.1 | 1 - 2 | 107 | 61 | |
NM_001028488.6 | NP_001023659.1 | 1 - 2 | 109 | 61 | ||
NM_001269507.4 | NP_001256436.1 | 1 - 2 | 113 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |