Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073799.5 | NP_506200.1 | 1 - 2 | 194 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |