Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047738.3 | NP_001041203.1 | 5 - 6 | 992 | 170 | |
NR_003178.1 | 5 - 6 | 957 | 170 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |