Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shw-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368471.3 | NP_001355445.1 | 8 - 9 | 1292 | 67 | |
NM_001380790.1 | NP_001368707.1 | 8 - 9 | 1283 | 67 | ||
NM_001380791.1 | NP_001366842.1 | 8 - 9 | 1317 | 67 | ||
GenBank mRNA | DQ185514.1 | - | 7 - 8 | 1229 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |