Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: him-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073876.4 | NP_506277.1 | 1 - 2 | 39 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |