Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zip-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129401.3 | NP_001122873.1 | 4 - 5 | 1397 | 91 | |
NM_073891.3 | NP_506292.1 | 3 - 4 | 475 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |