Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073906.2 | NP_506307.1 | 5 - 6 | 580 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |