Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bath-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073938.5 | NP_506339.2 | 2 - 3 | 691 | 154 |
Gene Symbol: F25H9.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073937.6 | NP_506338.1 | 6 - 7 | 1875 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |