Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: avr-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028905.8 | NP_001024076.1 | 3 - 4 | 705 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |