Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cav-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269594.2 | NP_001256523.1 | 3 - 4 | 350 | 110 | |
NM_001269596.3 | NP_001256525.1 | 2 - 3 | 183 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |