Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: letm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073980.6 | NP_506381.1 | 2 - 3 | 583 | 72 | |
NM_073981.7 | NP_506382.1 | 2 - 3 | 583 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |