Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074018.5 | NP_506419.3 | 3 - 4 | 390 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |