Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074190.3 | NP_506591.1 | 4 - 5 | 800 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |