Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269671.3 | NP_001256600.1 | 2 - 3 | 344 | 110 | |
NM_001269672.3 | NP_001256601.1 | 2 - 3 | 331 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |