Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269683.3 | NP_001256612.1 | 2 - 3 | 920 | 138 | |
NM_001269684.3 | NP_001256613.1 | 1 - 2 | 232 | 138 | ||
NM_001269685.3 | NP_001256614.1 | 1 - 2 | 232 | 138 | ||
NM_001276960.4 | NP_001263889.1 | 2 - 3 | 916 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |