Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scrm-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276962.2 | NP_001263891.1 | 4 - 5 | 906 | 110 | |
NM_074245.3 | NP_506646.1 | 3 - 4 | 717 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |