Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mut-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269709.2 | NP_001256638.1 | 6 - 7 | 1038 | 78 | |
NM_001269710.3 | NP_001256639.1 | 3 - 4 | 661 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |