Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269711.3 | NP_001256640.1 | 8 - 9 | 1495 | 67 | |
GenBank mRNA | AF318614.2 | - | 8 - 9 | 1471 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |