Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129445.5 | NP_001122917.1 | 5 - 6 | 543 | 149 | |
NM_001269713.3 | NP_001256642.1 | 1 - 2 | 57 | 149 | ||
NM_074294.5 | NP_506695.2 | 5 - 6 | 615 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |