Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cng-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129446.4 | NP_001122918.1 | 9 - 10 | 1943 | 92 | |
NR_070019.1 | 10 - 11 | 1738 | 92 | |||
NR_070020.1 | 9 - 10 | 1615 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |