Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269716.3 | NP_001256645.1 | 3 - 4 | 412 | 87 | |
NM_074311.6 | NP_506712.4 | 3 - 4 | 595 | 87 | ||
NM_171553.5 | NP_741641.2 | 3 - 4 | 409 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |