Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074385.4 | NP_506786.2 | 3 - 4 | 391 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |