Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074389.5 | NP_506790.1 | 2 - 3 | 774 | 84 | |
GenBank mRNA | L39890.1 | - | 2 - 3 | 750 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |